missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 506

Product Code. 15560607
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
100 tests
500 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15560607 100 Tests 100 tests
15570607 500 Tests 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15560607 Supplier Invitrogen™ Supplier No. 65086614
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation, and/or permeabilization procedures.

  • Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
  • Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
  • Fixable Viability Dye eFluor™ 506 can be excited by the violet (405 nm) laser line and has a peak emission of 506 nm that can be detected using a 510/50 band pass filter (equivalent to AmCyan)
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
  • After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
  • Fixable Viability Dye eFluor™ 506 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
  • It should be protected from light and moisture
  • Store at less than or equal to -70°C with dessicant, it may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
TRUSTED_SUSTAINABILITY

Specifications

Color Cyan
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
Emission 405/506
Product Line eBioscience
Quantity 100 Tests

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.