missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 450

Product Code. 15211938
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
Each
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15211938 100 Tests Each
15331988 500 Tests Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15211938 Supplier Invitrogen™ Supplier No. 65086314
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation, and/or permeabilization procedures.

  • Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
  • Thus, using these dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
  • Fixable Viability Dye eFluor™ 450 can be excited by the violet (405 nm) laser line and has a peak excitation of 450 nm that can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™)
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
  • After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
  • Fixable Viability Dye eFluor™ 450 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
  • It should be protected from light and moisture
  • Store at -70°C with dessicant, it may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
Excitation Wavelength Range 405/450
Product Line eBioscience
Quantity 100 Tests

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.