missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Ultra-Bright Key Chain Laser Pointer
Tous les produits Fisherbrand Produits
Click to view available options
:
Key Chain Laser Pointer
Description
Includes
Lithium battery powers unit for one year of intermittent use.
Designed specifically for slide presentations, wall charts, and field use.
- Beam diameter is 0.13 in. (3.5mm)
- With 4mW output and a range of 400 ft. (122m)
- Features key chain for easy portability
- Mini-light measures 2.50 x 1 x 0.25 in. dia. (6.4 x 2.5 x 0.64cm)
Specifications
Specifications
| Product Type | Laser Pointer |
| Wavelength | 650 nm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction