missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™

Product Code. 11724246 Shop All Fisherbrand Products
Change view
Click to view available options
Conductivity:
1 μS
10,000 μS
100 μS
100,000 μS
1000 μS
1413 μS
150,000 μS
200,000 μS
5 μS
Resistivity:
0.000005 μ℧
0.000006 μ℧
0.00001 mOhm
0.0001 mOhm
0.00071 mOhm
0.001 mOhm
0.01 mOhm
0.5 mOhm
1.0 mOhm
150000 μ℧
200000 μ℧
Concentration or Composition Notes:
NIST Batch Tested
NIST Individual Bottle Tested
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conductivity Resistivity Concentration or Composition Notes unitSize
11724246 1413 μS 0.00071 mOhm NIST Individual Bottle Tested Each
11754236 200,000 μS 0.000005 μ℧ NIST Individual Bottle Tested Each
11744236 150,000 μS 0.000006 μ℧ NIST Individual Bottle Tested Each
11784226 100,000 μS 0.00001 mOhm NIST Batch Tested Each
11704246 100,000 μS 0.00001 mOhm NIST Individual Bottle Tested Each
11774226 10,000 μS 0.0001 mOhm NIST Batch Tested Each
11794236 10,000 μS 0.0001 mOhm NIST Individual Bottle Tested Each
11714236 1413 μS 0.00071 mOhm NIST Batch Tested Each
11784236 1000 μS 0.001 mOhm NIST Individual Bottle Tested Each
11754226 100 μS 0.01 mOhm NIST Batch Tested Each
11774236 100 μS 0.01 mOhm NIST Individual Bottle Tested Each
11714246 5 μS 0.5 mOhm NIST Individual Bottle Tested Each
12399325 1 μS 1.0 mOhm NIST Individual Bottle Tested Each
11794226 150,000 μS 150000 μ℧ NIST Batch Tested Each
11704236 200,000 μS 200000 μ℧ NIST Batch Tested Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 11724246

Please to purchase this item. Need a web account? Register with us today!

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material

Order Info

473 mL (16 oz.) Bottle

Specifications

Chemical Name or Material Conductivity Standard
Description NIST Individual Bottle Tested, Resistivity: 0.00071 mΩ
Packaging Bottle
Concentration or Composition Notes NIST Individual Bottle Tested
Traceability to NIST NIST Individual Bottle Tested
Analyte or Component Names A2LA Conductivity Standards
Conductivity 1413 μS
Physical Form Liquid
Resistivity 0.00071 mOhm
Technique Calibration of Conductivity Instrument
Certifications/Compliance ISO 17034
Color Clear
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 1 x 473 mL (16 oz.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.