missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Shop All Fisherbrand ProductsDescription
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
- Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
- Also includes step-by step calibration instructions and temperature compensation chart
- Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
- ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Order Info
473 mL (16 oz.) Bottle
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Description | NIST Batch Tested, Resistivity: 200000 μ℧ |
| Packaging | Bottle |
| Concentration or Composition Notes | NIST Batch Tested |
| Traceability to NIST | NIST Batch Tested |
| Conductivity | 200,000 μS |
| Resistivity | 200000 μ℧ |
| Technique | Calibration of Conductivity Instrument |
| Certifications/Compliance | ISO 17034 |
| Color | Clear |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?