Learn More
Fisherbrand™ Stainless Steel Bow Tongs
Product Code. 11557872 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
150 mm
203 mm
Unit Size:
Each
Product Code. 11557872
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed using high quality components these corrosion resistant, non-magnetic bow tongs feature excellent heat resistance and great cryogenic stability.
  • 304 spec 18/8 non-magnetic stainless steel laboratory bow tongs with PTFE coated jaws
  • Corrosion resistant, non-magnetic bow tongs
  • Applied as a self priming single coat PTFE
  • Excellent heat resistance and great cryogenic stability
  • Excellent non-wetting properties that makes cleaning very easy and can also undergo steam or chemical sterilization
  • The coefficient of friction is generally in the range of 0.05 to 0.20
Specifications
Length (Metric) Opening 75 mm
Quantity 5 Pk.
Length (Metric) 203 mm
Material Stainless Steel (304)
Type Laboratory Bow Tongs