missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles

Catalog Number p-45803
Click to view available options
For Use With (Application):
Acetone
Ethanol
Isopropanol
Methanol
Unlabeled
This item is not returnable. View return policy
This item is not returnable. View return policy

Are pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.

  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

 

Use LYOB only with solvents compatible with LDPE.


Order Info

Prelabeled with six common solvents; also available unlabeled.

Capacity 16 oz., 500mL
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing Yes
Product Type Wash Bottle
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
For Use With (Application) Acetone
Material LDPE
Closure Color Red
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'