missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Silicone Recessed Septum Stoppers

Product Code. 11552503 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
13.75 to 14.25 mm
14.50 to 15.50 mm
20.00 to 21.00 mm
9.22 to 9.75 mm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
11552503 13.75 to 14.25 mm Pack of 10
11542503 9.22 to 9.75 mm Pack of 10
11572503 20.00 to 21.00 mm Pack of 10
11562503 14.50 to 15.50 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11552503
Only null left
Add to basket
Add to basket

Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal

  • Elliptical flange to allow efficient removal of stopper from aperature
  • Easily pierced with a hypodermic needle

Specifications

Diameter (Metric) Inner 13.75 to 14.25 mm
For Use With (Application) Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
For Use With (Equipment) For 13.75 to 14.25mm I.D. containers
Size 0
Color Translucent
Material Silicone
Product Type Septum Stopper
Stopper Number 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.