missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Labeled Wash Bottles

Product Code. p-45693 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
For Use With (Application):
Acetone
Deionized Water
Distilled Water
Isopropanol
Toluene
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 11512473

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Safety label includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Also includes Chemical Abstract Service (CAS) number, protective equipment symbols, and target organ information.

Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.

  • Easy-to-read printed bottles
  • Wide mouth 53 mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 mm, Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Use only with solvents compatible with LDPE

Order Info

Available in cases of six, all with the same label, or in a six-bottle assortment: one 500mL bottle each of acetone, ethanol, isopropanol, methanol, toluene and water.

Specifications

Capacity 17 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Natural
For Use With (Application) Isopropanol
Material Polyethylene
Sterility Non-sterile
Closure Color Yellow
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
Chemical Label Isopropanol
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.