missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Labeled Wash Bottles

Product Code. 11582463 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
For Use With (Application):
Acetone
Deionized Water
Distilled Water
Isopropanol
Toluene
Unit Size:
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) For Use With (Application) unitSize
11582463 1000 mL Acetone Pack of 6
11512473 500 mL Isopropanol Pack of 6
10022564 500 mL Acetone Pack of 6
11522473 500 mL Toluene Pack of 6
11532473 500 mL Deionized Water Pack of 6
11562463 500 mL Distilled Water Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11582463

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Safety label includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Also includes Chemical Abstract Service (CAS) number, protective equipment symbols, and target organ information.

Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.

  • Easy-to-read printed bottles
  • Wide mouth 53 mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 mm, Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Use only with solvents compatible with LDPE

Order Info

Available in cases of six, all with the same label, or in a six-bottle assortment: one 500mL bottle each of acetone, ethanol, isopropanol, methanol, toluene and water.

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Natural
For Use With (Application) Acetone
Material Polyethylene
Sterility Non-sterile
Closure Color Red
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
Chemical Label Acetone
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.