Learn More
Fisherbrand™ SafeSnap™ Disposable Ampule Openers
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
1 to 4mL ampule
Unit Size:
Pack of 1000
Description
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
- To use: Slip the collar onto the ampule until collar rests on the widest part
- Grip the collar with thumb and forefinger while holding the upright ampule at bottom
- Break the ampule with a quick snap
- Dispose of the collar and ampule top together
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Ampule Opener |
| Material | Polyethylene |
| For Use With (Equipment) | 1 to 4mL ampule |