Description
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
- To use: Slip the collar onto the ampule until collar rests on the widest part
- Grip the collar with thumb and forefinger while holding the upright ampule at bottom
- Break the ampule with a quick snap
- Dispose of the collar and ampule top together
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Ampule Opener |
| Material | Polyethylene |
| For Use With (Equipment) | 1 to 4mL ampule |