missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rubber-Lined Plastic Caps

Product Code. 11518222 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
22-400
38-400
58-400
Unit Size:
Pack of 288
Pack of 576
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish unitSize
11518222 58-400 Pack of 288
11728709 22-400 Pack of 576
11588212 38-400 Pack of 576
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11518222

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Resilient, resistant to moisture vapor

  • Black, rubber-lined caps Autoclavable
  • Packaged in 12 packs

Specifications

Color Black
Description Screw cap
Material Plastic
No. per Case 288 Cs.
Quantity 288/PK.
Liner Material Rubber
Neck Finish 58-400
Certifications/Compliance ISO 9001
Product Type Screw Cap
For Use With (Equipment) Bottle
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.