missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Click to view available options
Capacity (Metric):
15 mL
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
Beschreibung
Rib construction permits stacking without binding
- Special “dripless” lip
- Base allows beaker to sit flat during use
- Graduated in mL and oz.
Spezifikation
Capacity (Metric) | 150 mL |
Material | Polypropylene |
For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
Includes | Spout |
Increments | 5 mL |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Single-use |
Graduated | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'