missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Product Code. 18386513 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
Clear with Red Print
Natural
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
24 x 36 in. (61 x 91 cm)
25 x 35 in. (64 x 89 cm)
30 x 24 in. (76 x 61 cm)
30 x 33 in. (76 x 84 cm)
36 x 24 in. (91 x 61 cm)
48 x 38 in. (122 x 97 cm)
Thickness:
1.50 mil
4 mil
Unit Size:
Each
Pack of 100
Pack of 200
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Thickness unitSize
18386513 Red with Black Print 30 x 33 in. (76 x 84 cm) 1.50 mil Pack of 200
15386433 Natural 24 x 12 in. (61 x 30 cm) 1.50 mil Pack of 200
16235040 Natural 24 x 36 in. (61 x 91 cm) 1.50 mil Each
18346523 Clear with Red Print 12 x 8 in. (30 x 20 cm) 1.50 mil Pack of 200
18396533 Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil Pack of 200
18366533 Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil Pack of 200
18366513 Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil Pack of 200
18376513 Red with Black Print 12 x 8 in. (30 x 20 cm) 1.50 mil Pack of 200
18376523 Red with Black Print 19 x 14 in. (48 x 36 cm) 1.50 mil Pack of 200
17841772 Red with Black Print 23 x 19 in. (58 x 48 cm) 1.50 mil Pack of 200
18396523 Red with Black Print 25 x 35 in. (64 x 89 cm) 1.50 mil Pack of 200
18316513 Red with Black Print 36 x 24 in. (91 x 61 cm) 4 mil Pack of 100
18346533 Clear 36 x 24 in. (91 x 61 cm) 4 mil Pack of 100
18346513 Clear 30 x 24 in. (76 x 61 cm) 1.50 mil Pack of 200
18548056 Red with Black Print 48 x 38 in. (122 x 97 cm) 1.50 mil Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 18386513
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles

Specifications

Certifications/Compliance OSHA, 29 CFR1910.1030
Closure Type Elastic
Color Red with Black Print
Dimensions (L x W) 30 x 33 in. (76 x 84 cm)
Length (Metric) 76 cm
Width (Metric) 84 cm
Autoclavable Autoclavable
Includes Printed in English and Spanish
Width (English) 33 in.
Length (English) 30 in.
Thickness 1.50 mil
Material Polyethylene
Product Type Autoclavable Bags
Temperature (Metric) 127°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.