missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
Specifications
| Certifications/Compliance | OSHA, 29 CFR1910.1030 |
| Closure Type | Elastic |
| Color | Red with Black Print |
| Dimensions (L x W) | 25 x 35 in. (64 x 89 cm) |
| Length (Metric) | 63.5 cm |
| Width (Metric) | 88.9 cm |
| Autoclavable | Autoclavable |
| Includes | Printed in English and Spanish |
| Width (English) | 35 in. |
| Length (English) | 25 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?