missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Test Paper Rolls

Product Code. 17785738 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0 to 14
Quantity:
100 Tests/Roll, 700 Total; Set of each pH range dispenser (7)
Unit Size:
Pack of 700
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Quantity Unit Size
17785738 0 to 14 100 Tests/Roll, 700 Total; Set of each pH range dispenser (7) Pack of 700
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17785738
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

General purpose pH papers in efficient 15 ft. rolls

  • Convenient dispenser allows you to select the right amount of pH paper for each application
  • Easy-to-use—Dip paper in solution to be tested and match to color chart immediately
  • Highly accurate and well suited for most applications
  • Each SKU is packed in box of five clear plastic dispensers
  • Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)

Order Info

15 ft. Roll provides about 100 tests.

Specifications

pH Range 0 to 14
Format Strip
For Use With (Application) Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications
Includes 13640500, 13640501, 13640502, 13640503, 13640504, 13640505, 13640506
Quantity 100 Tests/Roll, 700 Total; Set of each pH range dispenser (7)
Storage Requirements Room Temperature
Type pH Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.