missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PE Cone-Lined Polyethylene Caps

Product Code. 11518182 Shop All Fisherbrand Products
Change view
Click to view available options
Unit Size:
Pack of 2300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11518182 Pack of 2300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11518182

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Black
Description Screw cap
Material Phenolic Resin
Length (Metric) 36mm
No. per Case 2300 Cs.
Quantity 2200/Cs.
Liner Material PolyCone
Neck Finish 33-400
Certifications/Compliance ISO 9001
Shape Round
Product Type PE Cone Lined Cap
For Use With (Equipment) Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.