missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Handheld Digital Clinical Refractometer

Product Code. 18820083 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Refractive Index:
1.3330 to 1.4200
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index Unit Size
18820083 1.3330 to 1.4200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18820083
  • ONLINE EXCLUSIVE PRICE
    £1686.83
    £932.89 / Pack of 1
    Save £753.94
    44% Off
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Digital refractometer, Rubber grip, prism cover, hard shell case, battery, Applicator, Manual

Fisherbrand™ Handheld Digital Clinical Refractometers are portable, reliable and accurate for fast, reliable results to determine dehydration or total plasma protein levels in blood.

Fisherbrand™ Digital handheld Refractometers are designed to fit into the palm of your hands, and easy to operate. Their compact design allows them to be taken anywhere you need them to go with its IP65 ingress protection water and dust. .

Each unit is inspected in the United States by a factory qualified technician under a ridged quality control process to insure its accuracy, and performance.

KEY FEATURES:

  • Compact, lightweight design for easy portability
  • Automatic Temperature Compensation to prevent inaccurate readings due to temperature fulgurations.
  • Stainless steel prism well for great protection and easy cleaning
  • Easily switch between C°/ F° readings
  • IP65 rating for protection against water and dust
  • 589.3nm LED for easy readings in any low light situation, and increased accuracyPowered by a single AAA battery
  • Battery-saving mode automatically powers unit off after 1 minute of inactivity
  • Requires 4 to 5 drops sample size for micro-volume detection
  • AFM technology measures the sample 15 times, and displays the mean value
  • Large digital display, allows for easy-to-read measured values, temperature, and battery life all on the display
  • Removable grip for easy cleaning
  • Hanging strap to prevent accidentally dropping the unit
  • Single button calibration with distilled water
  • 4 rubber feet allow for the unit to sit firmly and securely on most surfaces without causing scratches to the unit or the surface it is sitting on
  • 1-Year warranty

 

CLINICAL MODELS

Quickly and accurately get an indication of basic health parameters such as dehydration and blood plasma protein levels FisherBrand™ Handheld Digital Clinical Refractometers measures Total Serum Protein, Urine Specific Gravity, and Refractive Index, and ideal for use in quality control, research, and clinical laboratories.

Warranty

1 year

Specifications

Type Clinical Refractometer
Scale Range -0.1 to 12.0 Serum Protein, 1.000 to 1.050 Urine Specific Gravity
Refractive Index 1.3330 to 1.4200
Sample Volume >0.2 mL
Calibration Distilled water
Accuracy Urine SP. G. ±0.002; Serum P. ±0.2; Refractive Index ±0.0003nD
Resolution SP. G. 0.001; S.P. 0.1; RI 0.0001nD
Display LCD
Increments SP. G. 0.001; S.P. 0.1; RI 0.0001nD
Scale Type Linear
Measurement Time 2 sec.
Lighting Type LED
For Use With (Application) Clinical; Veterinary; Drug testing; Dehydration
Includes Digital refractometer, Rubber grip, Prism cover, Hard shell case, Battery, Applicator, Manual
IP Rating IP65
Quantity 1
Weight (English) 0.2 lb.
Weight (Metric) 90 g
Dimensions (L x W x H) 25 x 58 x 121 mm
Height (English) 0.98 in.
Height (Metric) 121 mm
Length (English) 4.76 in.
Length (Metric) 25 mm
Width (English) 2.28 in.
Width (Metric) 58 mm
Temperature Compensation Yes
Temperature Range (Metric) 0°C to 40°C
Temperature Resolution 0.1°C
Power Supply Details 1xAAA Battery
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.