missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Flint Glass Tubes with Plain End

Specially annealed to reduce breakage along stress point

Brand:  Fisherbrand™

Code : 40

Additional Details : Weight : 2.64000kg

 View more versions of this product

Product Code. 15135134

  • £110.00 / Pack of 1000


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Round bottom
  • Smooth top and bottom to prevent cuts

Specifications

Specifications

15mm
Specially annealed to reduce breakage along stress point
Tray
11mL
85mm
Disposable Culture Tube
No
Yes
None
250/Tray
KIM-KAP
Flint Glass
Round Bottom
Videos