Learn More
Fisherbrand™ Compartmentalized Petri Dishes
Product Code. 16123707 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Tri Dish, Y-Plate
Unit Size:
Pack of 600
Product Code. Type unitSize
16123707 Tri Dish, Y-Plate Pack of 600
1 options
Product Code. 16123707
Only null left
Add to basket
Add to basket
Select from 2, 3, or 4-section plates with numbered sections
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Closure Type Stackable
Material Polystyrene
Sterility Sterile
Type Tri Dish, Y-Plate
Edges Raised Ridge
Packaging 24/Sleeve, 25 Sleeves/Cs.
Height (Metric) 15 mm
Diameter (Metric) 65 mm