missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 2mm Thick
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Descripción
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximises sample volume
- 18-sample and 36-sample combs are Multichannel pipettor compatible
Especificaciones
Blue | |
Comb | |
1 Each | |
400 μL |
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units | |
5 | |
2 mm |