missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 1.5mm Thick

For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11807843

  • £21.70 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Red
Comb
1 Each
320 μL
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
5
1.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers