missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 0.75mm Thick

For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11847833

  • £20.30 / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Black
Comb
1 Each
15 μL
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
48
0.75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers