missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 1.5mm Thick

For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11897553

  • £27.20 / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Red
Comb
1 Each
72 μL
Sub-Gel Mini Horizontal Gel Electrophoresis Unit
4 Prep, 2 Marker
1.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers