missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 1.5mm Thick
For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximises sample volume
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications
Red | |
Comb | |
1 Each | |
20 μL |
Sub-Gel Mini Horizontal Gel Electrophoresis Unit | |
12 | |
1.5 mm |