missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 0.75mm Thick

For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11857563

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Black
Comb
1 Each
8 μL
Sub-Gel Mini Horizontal Gel Electrophoresis Unit
8
0.75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers