missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 1mm Thick

Product Code. 11553362 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
4
8
10
12
16
20
26
Volume (Metric) Sample:
22 μL
29 μL
40 μL
42 μL
52 μL
67 μL
Unit Size:
Each
Product Code. No. of Wells Volume (Metric) Sample unitSize
11553362 12 40 μL Each
11543362 10 52 μL Each
11573362 20 22 μL Each
11563362 16 29 μL Each
11523362 4 42 μL Each
11533362 8 67 μL Each
11583362 26 - Each
7 options

Product Code. 11553362

Only null left
Add to basket
Add to basket

For use with Mini-Plus Wide Format Horizontal Gel Units.

  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 16-sample combs are Multichannel pipettor compatible

Specifications

Color White
For Use With (Equipment) Mini-Plus Wide Format Horizontal Gel Units
Type Comb
No. of Wells 12
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Sample 40 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.