Learn More
Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 1.5mm Thick
Product Code. 11503372 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
4
8
10
12
16
20
26
Volume (Metric) Sample:
26 μL
32 μL
44 μL
61 μL
77 μL
100 μL
213 μL
Unit Size:
Each
Product Code. 11503372
Only null left
Add to basket
Add to basket
For use with Mini-Plus Wide Format Horizontal Gel Units.
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
For Use With (Equipment) Mini-Plus Wide Format Horizontal Gel Units
Type Comb
No. of Wells 4
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 213 μL