missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Midi-Plus Wide Format Horizontal Gel Units, 2mm Thick

Product Code. 11503412 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
12
16
20
25
28
40
50
Volume (Metric) Sample:
30 μL
34 μL
52 μL
60 μL
80 μL
104 μL
144 μL
Unit Size:
Each

Product Code. 11503412

Only null left
Add to basket
Add to basket

For use with Midi-Plus Wide Format Horizontal Gel Units.

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 12-sample, 25-sample and 50-sample combs are Multichannel pipettor compatible

Specifications

Color Blue
For Use With (Equipment) Midi-Plus Wide Format Horizontal Gel Units
Type Comb
No. of Wells 28
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Sample 52 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.