missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Midi-Plus Wide Format Horizontal Gel Units, 1mm Thick

Product Code. 11583392 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
12
16
20
25
28
40
50
Volume (Metric) Sample:
15 μL
17 μL
26 μL
30 μL
40 μL
52 μL
72 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11583392 50 15 μL Each
11523392 12 72 μL Each
11533392 16 52 μL Each
11543392 20 40 μL Each
11553392 25 30 μL Each
11563392 28 26 μL Each
11573392 40 17 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11583392
Only null left
Add to basket
Add to basket

For use with Midi-Plus Wide Format Horizontal Gel Units.

  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 12-sample, 25-sample and 50-sample combs are Multichannel pipettor compatible

Specifications

Color White
For Use With (Equipment) Midi-Plus Wide Format Horizontal Gel Units
Type Comb
No. of Wells 50
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Sample 15 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.