missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-Coded Autoclavable Identification Tape

Product Code. 17837261 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Red
Silver
Yellow
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17837261 Yellow Each
17867261 Orange Each
17857261 Red Each
17847261 Green Each
17877261 Blue Each
17887261 Brown Each
17807271 Black Each
17897261 Silver Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17837261
Only null left
Add to basket
Add to basket

Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.

  • Vinyl self-stick tapes color-code autoclave items such as surgical instruments for easy identification
  • Attach securely and safely to any clean surface
  • Resist water, oil, and acids
  • Nontoxic

Order Info

In 16 ft.L x 0.37 in. W (5m x 9.6mm) rolls.

Specifications

Autoclavable Autoclavable
Color Yellow
Format Tape
Material Vinyl
Product Type Identification Tape
For Use With (Application) Resist Water, Oil and Acids
Quantity 1 Ea.
Length (English) 16 ft.
Length (Metric) 4.8 m
Width (English) 0.37 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.