missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls

Product Code. 16237040 Shop All Fisherbrand Products
Click to view available options
Product Type:
Biohazard Liner
Biohazard Roll
Biohazard Wipe
Biohazard Wipes
Absorbancy:
Heavy
Standard
Super
Dimensions (L x W):
100 ft. x 16 in.
16 x 16 in.
22 x 16 in.
3 x 3 in. (7.62 x 7.62 cm)
4 x 4 in. (10.16 x 10.16 cm)
9 x 8 in. (22.86 x 20.32 cm)
missing translation for 'unitSize'
Each
Pack of 100
Pack of 125
Pack of 200
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 16237040

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prevent leakage of fluids and chemicals to hands, gloves and work surfaces

Wipes and Liners are non-sterile and not intended for use on patients.

  • Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
  • Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
  • Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
  • Can also be used as mats and liners; rolls can be cut to desired length
  • One time use only

Specifications

Product Type Biohazard Roll
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 16 in.
Width (Metric) 40.64 cm
Material Non-woven Plastic Cellulose
Absorbancy Super
Color White, Orange
Disposable Single-use
Dimensions (L x W) 100 ft. x 16 in.
For Use With (Application) Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates.
For Use With (Equipment) Bench tops, specimen collection tubs, trays, fume hoods, Bio-Safety and chemical cabinets
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.