missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Tall-Chimney PCR Plate

Product Code. 15265143 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15265143 Natural Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15265143
Only null left
Add to basket
Add to basket

Compatible with all leading thermal cyclers and MegaBACE™ capillary sequencers†

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Highly rigid plate deck
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 340μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumic grid
  • Cleanroom produced

Specifications

Format Standard
Array 8 x 12
Color Natural
Product Type PCR Microplate
Skirt Style Non-skirt
Material Polypropylene
Volume (Metric) Well 340 μL
No. of Wells 96
For Use With (Application) PCR
Rimmed Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.