missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Virgin polypropylene
- Rigid plate deck
- 40μL maximum well volume; 25μL working volume
- Cut-away corners and alphanumeric grid
- SBS (Society for Biomolecular Screening) footprint
- Automation friendly
- Eight holes in skirt aid plate positioning and removal from thermal cycler block
- Cleanroom produced
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | Natural |
| For Use With (Equipment) | 384-block PCR and sequencing instruments |
| Material | Polypropylene |
| No. of Wells | 384 |
| Volume (Metric) Working | 25 μL |
| Rimmed | Yes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?