missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polystyrene Plates

Product Code. p-153498 Shop All Fisherbrand Products
Click to view available options
Color:
Black
Clear
White
Well Shape:
Rounded Square
Triangular, Flat Bottom (F-well)
Unit Size:
Pack of 100
Pack of 90

Product Code. 11947954

Only null left
Add to basket
Add to basket

Process four times as many samples as standard 96-well plates

Optimized for robotic instruments, plate readers and liquid handling systems.

  • Highly specified polystyrene
  • Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
  • Flat well bottom surface provides greater area for binding
  • Working volume: 10–100μL
  • 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
  • Untreated
  • Non-sterile

Order Info

Accommodate 12-565-511 lid (sold separately).

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Rounded Square
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.