missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Optimized for robotic instruments, plate readers and liquid handling systems.
- Highly specified polystyrene
- Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
- Flat well bottom surface provides greater area for binding
- Working volume: 10–100μL
- 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
- Untreated
- Non-sterile
Order Info
Accommodate 12-565-511 lid (sold separately).
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 10 to 100 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Rounded Square |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?