missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 8-Tooth Combs for Horizontal Midigel Systems

8-Tooth Combs for Horizontal Midigel Systems

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.22000kg

 View more versions of this product

Product Code. 12483637

  • £46.65 / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

  • Accessory for use with Horizontal Midigel Systems

Specifications

Specifications

Owl™ A1 Large Gel Electrophoresis System
16 x 13 cm
0.1 cm
0.54 in.
Standard Comb
8
65 μL
1.39 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers