missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 12-Tooth Combs for Horizontal Midigel Systems

12-Tooth Combs for Horizontal Midigel Systems

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.81650kg

 View more versions of this product

Product Code. 12413637

  • £46.65 / Each
Estimated Shipment: 09-07-2024
to see stock.
Add to basket

Description

Description

  • Accessory for use with Horizontal Midigel Systems

Specifications

Specifications

Owl A1 Large Gel Electrophoresis System
12
40 μL
0.87 cm
Standard Comb
0.1 cm
0.34 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers