Learn More
Fisherbrand™ Vortex Mixer Accessory, Insert Retainer
Keeps tube holder in place when used with vortex mixer.
Brand:  Fisherbrand™
Additional Details : Weight : 0.09000kg
Product Code. 15766939
  • £130.03 / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
Keeps tube holder in place when used with vortex mixer.
  • Required to secure foam inserts or vessel harness
  • Compatible with other manufacturers' models(contact your Fisher Scientific Customer Service Representative for details)
Specifications
Specifications
Insert Retainer
Vortex Mixer Foam Insert