Learn More
Fisherbrand™ Vortex Mixer Head
Product Code. 15756949 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Vortex mixers
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15756949 Vortex mixers Each
1 options
Product Code. 15756949
Only null left
Add to basket
Add to basket
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
TRUSTED_SUSTAINABILITY
Specifications
Product Type Head
Diameter (Metric) 76 mm
For Use With (Equipment) Vortex mixers