missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vortex Mixer Accessories: Foam Insert Kit

For use with Fisherbrand vortex mixers.

Brand:  Fisherbrand™

Additional Details : Weight : 0.16000kg

 View more versions of this product

Product Code. 15776939

  • £74.20 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.

Foam Insert Variety Pack

  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm

Specifications

Specifications

Foam Insert Kit
Vortex mixer
9 to 13mm Tube holder, 4 to 19mm tube holder, 20 to 29mm tube holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers