missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometer

Product Code. p-3742835 Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
-2.00°C to +2.00°C
-50°C to +70°C
23.00°C to 27.00°C
35°C to 39°C
Temperature Range (English):
-58° to +158°F
28.4°F to 35.6°F
37.00° F
73.4°F to 80.6°F
Unit Size:
Each

Product Code. 11749705

Only null left
Add to basket
Add to basket

Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.

Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.

  • Provides accurate readings with tip penetration of 0.33 in.
  • Records time/date for min/max readings and hi/lo alarm alerts
  • Displays minimum/maximum and current temperatures
  • Min/max monitors readings over any period; displays exact time/date of min/max temperature
  • Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
  • Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
  • Resolution: 0.01°
  • Accuracy: ±0.05°C within ±2°C of standard
  • Probe Dia.: 0.12 in. (0.30 cm)
  • Stem Length: 6.25 in. (15.87 cm)
  • Overall Length: 9 in. (22.86 cm)
  • Cable: 5 ft. (1.5 m)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Compliance

Meets ITS-90, International Temperature Standard.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.12 in.
Diameter (Metric) Probe 3 mm
Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9 cm)
For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
Height (English) 0.75 in.
Height (Metric) 1.9 cm
Includes Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)
Length (English) Probe 6.25 in.
Length (Metric) 107 mm
Length (Metric) Probe 15.9 cm
Material ABS Plastic
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 140 g
Width (English) 2.75 in.
Product Type Digital Thermometer
Length (English) 4.25 in.
Max. Temperature (English) 35.6°F
Max. Temperature (Metric) 2°C
Min. Temperature (English) 28.4°F
Min. Temperature (Metric) -2°C
Temperature Range (English) 28.4°F to 35.6°F
Temperature Range (Metric) -2.00°C to +2.00°C
Calibrated Yes
For Use With (Equipment) Water Baths
Battery Type AA
Accuracy ±0.05°C
Display LED
Width (Metric) 6.9 cm
Scale Reads to 0.00
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.