missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fisher Scientific™ Traceable™ Flip-Stick™ Thermometer with fold-out design makes taking probe readings in any position a snap.
- Features °F/°C switch
- Stainless-steel probe
- Shockproof and drop-proof up to 5 ft.
- Max/min temperature reading over any period
- Wide temperature range cover most routine measurements
- Position probe at any angle
- Protect probe in fold-up position
- 0.75 in. Display is easy to read
- Rapid reading update—every second
- HOLD switch freezes display to capture a temperature
Certifications
Individually serial-numbered Traceable Certificate is provided from ISO 17025 calibration laboratory accredited by A2LA indicating traceability to standards provided by NIST (National Institute of Standards and Technology).
Specifications
Specifications
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Dimensions (L x W x H) | 11 x 0.6 in. (28 x 1.5 cm) |
| For Use With (Application) | Food, gases, samples, reagents, water baths and semi solids |
| Includes | Stainless Steel probe |
| Length (English) Probe | 4.5 in. |
| Length (Metric) | 280 mm |
| Length (Metric) Probe | 11.4 cm |
| Material | Plastic |
| Resolution | 0.1°C |
| Weight (English) | 2.4 oz. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?