Learn More
Fisherbrand™ Traceable™ DAS-4 Data Logger System
Product Code. 11784276 Shop All Fisherbrand Products
Click to view available options
Data Memory:
8000 readings
Unit Size:
Each
Product Code. 11784276
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Captures and stores up to 8000 readings
  • Readings may be taken at intervals from 1 second to 99 hours
  • Stored readings may be downloaded to any IBM-compatible computer
  • Data files can be read by spreadsheets, databases, and statistical programs
Order Info
Supplied with 36 in. (91.4cm) serial cable with D9F plug, 3.50 in. diskette, and four AA alkaline batteries. Unit measures 1L x 3W x 5 in. H (2.5 x 7.6 x 12.7cm); weighs 7 oz. (198.4g)
Specifications
Type Datalogger System
Data Memory 8000 readings
Calibration Not Calibrated
For Use With (Application) Captures and stores up to 8000 readings