missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™

Product Code. 15598693 Shop All Fisherbrand Products
Change view
Click to view available options
pH:
4.005
7.000
10.012
Color:
Blue
Red
Yellow
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Color unitSize
15598693 4.005 Red 500mL
15508703 7.000 Yellow 500mL
15518703 10.012 Blue 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15598693

Please to purchase this item. Need a web account? Register with us today!

Edge

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.

  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Each 16 ounce bottle or six-pack single-use 100ml containers is supplied with individual temperature chart, traceability information and Traceable™ Certificate

Specifications

Chemical Name or Material Buffer Standards pH 4.005(CRM)
Color Red
For Use With (Application) Calibration
For Use With (Equipment) pH Instruments and Electrodes
pH 4.005
Physical Form Liquid
Quantity 473 mL
Solution Type Buffer
Packaging 473mL (16 oz.) bottle (ea)
Traceability to NIST 'Certified and Traceable™ ' ‘ISO 17034 (formerly ISO Guide 34)’ and ‘Certified Reference Material’
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.