missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ One-Shot™ pH Buffer Standards pH Certified Reference Material (CRM), Fisherbrand™
Shop All Fisherbrand ProductsDescription
Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied with individual temperature chart, traceability information and Traceable™ Certificate
Specifications
Specifications
| Chemical Name or Material | Buffer Standards One-Shot (CRM) |
| Color | Blue |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 10.012 |
| Physical Form | Liquid |
| Quantity | 6 x 100 mL |
| Solution Type | Buffer |
| Packaging | 100 mL One-Shot (6 Pk.) |
| Traceability to NIST | Certified and Traceable™ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?