Learn More
Fisherbrand™ Maxima™ C Plus Pumps, Standard Exhaust Oil Filter Replacement Elements
Replacement filter will help keep the Macima C vacuum pump running smoothly
Brand: Fisherbrand™
Code : 45
Additional Details : Weight : 1.00000kg
Description
Replacement filter will help keep the Macima C vacuum pump running smoothly
For exhaust filters supplied with Fisher Scientific Maxima C Plus pumps.
Specifications
Model M12C and M16C vacuum pump exhaust filter | |
Filter Element |