Learn More
Thermo Scientific™ Thermo Scientifi™ Chiller External Pressure Reducers
Product Code. 11987514 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
ThermoChill I and II
ThermoChill III
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11987514 ThermoChill III Each
11977514 ThermoChill I and II Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11987514 Supplier Thermo Scientific™ Supplier No. 196000000001
External pressure reducers
External pressure reducers for ThermoChill Recirculating chillers
  • 1/2 in. external pressure reducer
  • Adjustable to safely reduce or limit system pressure and flow
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoChill III
Type External Pressure Reducer (EPR)
Size 1/2 in.