missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Circular Bottom Desi-Vac™ Container Desiccators with Vacuum Pump

No separate vacuum pump is required for these desiccators.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11764366

  • £91.60 / Each


In Stock 44
Add to basket

Description

Description

Plastic container with a self-contained vacuum pump is ideal for drying, storing chemicals, powders, sample tissues and plant sections, etc.

  • Self-contained unit holds a vacuum of 14.7 in.Hg (373mm) in excess of 24 hours
  • Vacuum pump built into the top removes air with manual strokes
  • Rubber O-ring sealed cover maintains vacuum
  • Release button on top allows air into the container to remove the lid
  • Use with Fisherbrand™ Humidity Sponge™ to keep samples dry

Specifications

Specifications

Circular
Polycarbonate
Desiccator
21.5cm
Metal pump handle, chemical-resistant ABS plastic vacuum pump and cover, release button on top, O-Ring-sealed cover
1500cc
18.4cm
Videos
Documentation